accessibility menu, dialog, popup

UserName

FOO

CAS RN 68359-57-9

IUPAC Name: 4-fluoro-3-phenoxybenzaldehyde
Synonyms: 4-fluoro-3-phenoxybenzaldehyde
Molecular Weight (g/mol): 216.21
Molecular Formula: C13H9FO2
InChi Key: JDICMOLUAHZVDS-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=O)C=C1OC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results