missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-3-nitrobenzotrifluoride 95.0+%, TCI America™
$42.87 - $197.59
Chemical Identifiers
| CAS | 367-86-2 |
|---|---|
| Molecular Formula | C7H3F4NO2 |
| Molecular Weight (g/mol) | 209.1 |
| MDL Number | MFCD00007059 |
| InChI Key | HLDFCCHSOZWKAA-UHFFFAOYSA-N |
| Synonym | 4-fluoro-3-nitrobenzotrifluoride, 1-fluoro-2-nitro-4-trifluoromethyl benzene, benzene, 1-fluoro-2-nitro-4-trifluoromethyl, 4-fluoro-3-nitrotrifluoromethylbenzene, 3-nitro-4-fluorobenzotrifluoride, 3-nitro-alpha,alpha,alpha,4-tetrafluorotoluene, 2-fluoro-5-trifluoromethyl nitrobenzene, 3-nitro-4-fluoro benzotrifluoride, 1-fluoro-2-nitro-4-trifluoromethyl-benzene |
| PubChem CID | 67778 |
| IUPAC Name | 1-fluoro-2-nitro-4-(trifluoromethyl)benzene |
| SMILES | C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])F |
Chemical Identifiers
| 367-86-2 | |
| 209.1 | |
| HLDFCCHSOZWKAA-UHFFFAOYSA-N | |
| 67778 | |
| C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])F |
| C7H3F4NO2 | |
| MFCD00007059 | |
| 4-fluoro-3-nitrobenzotrifluoride, 1-fluoro-2-nitro-4-trifluoromethyl benzene, benzene, 1-fluoro-2-nitro-4-trifluoromethyl, 4-fluoro-3-nitrotrifluoromethylbenzene, 3-nitro-4-fluorobenzotrifluoride, 3-nitro-alpha,alpha,alpha,4-tetrafluorotoluene, 2-fluoro-5-trifluoromethyl nitrobenzene, 3-nitro-4-fluoro benzotrifluoride, 1-fluoro-2-nitro-4-trifluoromethyl-benzene | |
| 1-fluoro-2-nitro-4-(trifluoromethyl)benzene |
Specifications
| 367-86-2 | |
| C7H3F4NO2 | |
| 25 g | |
| 4-fluoro-3-nitrobenzotrifluoride, 1-fluoro-2-nitro-4-trifluoromethyl benzene, benzene, 1-fluoro-2-nitro-4-trifluoromethyl, 4-fluoro-3-nitrotrifluoromethylbenzene, 3-nitro-4-fluorobenzotrifluoride, 3-nitro-alpha,alpha,alpha,4-tetrafluorotoluene, 2-fluoro-5-trifluoromethyl nitrobenzene, 3-nitro-4-fluoro benzotrifluoride, 1-fluoro-2-nitro-4-trifluoromethyl-benzene | |
| C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])F | |
| 209.1 | |
| 209.10 | |
| Liquid |
| 208°C | |
| MFCD00007059 | |
| 1993 | |
| HLDFCCHSOZWKAA-UHFFFAOYSA-N | |
| 1-fluoro-2-nitro-4-(trifluoromethyl)benzene | |
| 67778 | |
| ≥95.0% (GC) | |
| 4-Fluoro-3-nitrobenzotrifluoride |
Safety and Handling
TSCA : No