accessibility menu, dialog, popup

UserName

CAS RN 367-86-2

IUPAC Name: 1-fluoro-2-nitro-4-(trifluoromethyl)benzene
Synonyms: 1-fluoro-2-nitro-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 209.1
Molecular Formula: C7H3F4NO2
InChi Key: HLDFCCHSOZWKAA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC=C1F)C(F)(F)F
Boiling Point 
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)