Learn More
4-Fluoro-3-nitrobenzotrifluoride, 97%
CAS: 367-86-2 | C7H3F4NO2 | 209.1 g/mol
$56.49 - $458.97
Chemical Identifiers
| CAS | 367-86-2 |
|---|---|
| Molecular Formula | C7H3F4NO2 |
| Molecular Weight (g/mol) | 209.1 |
| MDL Number | MFCD00007059 |
| InChI Key | HLDFCCHSOZWKAA-UHFFFAOYSA-N |
| Synonym | 4-fluoro-3-nitrobenzotrifluoride, 1-fluoro-2-nitro-4-trifluoromethyl benzene, benzene, 1-fluoro-2-nitro-4-trifluoromethyl, 4-fluoro-3-nitrotrifluoromethylbenzene, 3-nitro-4-fluorobenzotrifluoride, 3-nitro-alpha,alpha,alpha,4-tetrafluorotoluene, 2-fluoro-5-trifluoromethyl nitrobenzene, 3-nitro-4-fluoro benzotrifluoride, 1-fluoro-2-nitro-4-trifluoromethyl-benzene |
| PubChem CID | 67778 |
| IUPAC Name | 1-fluoro-2-nitro-4-(trifluoromethyl)benzene |
| SMILES | C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1967614
|
Thermo Scientific Chemicals
A1967614 |
25 g |
Each for $56.49
|
|
|||||
|
AAA1967622
|
Thermo Scientific Chemicals
A1967622 |
100 g |
Each for $138.22
|
|
|||||
|
AAA1967636
|
Thermo Scientific Chemicals
A1967636 |
500 g |
Each for $458.97
|
|
|||||
Description
4-Fluoro-3-nitrobenzotrifluoride is used in the pre-column derivatization technique for HPLC with UV/VIS spectrophotometric detection of polyamines. It may be used as derivatization reagent for the HPLC determination of polyamines. It was also used in the synthesis of 2-nitro-4-trifluoromethylphenylhydrazine, derivatization reagent for 1,2-diol compounds and oxo-steroids.
- [3-(2-nitro-4-trifluoromethylphenyl)aminophenyl]dihydroxyborane, derivatization reagent for 1,2-diol compounds and oxo-steroids3
- 1-(3-chlorophenyl)-5-trifluoromethyl-3-hydrobenzimidazol-2-one
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Fluoro-3-nitrobenzotrifluoride is used in the pre-column derivatization technique for HPLC with UV/VIS spectrophotometric detection of polyamines. It may be used as derivatization reagent for the HPLC determination of polyamines. It was also used in the synthesis of 2-nitro-4-trifluoromethylphenylhydrazine, derivatization reagent for 1,2-diol compounds and oxo-steroids. • [3-(2-nitro-4-trifluoromethylphenyl)aminophenyl]dihydroxyborane, derivatization reagent for 1,2-diol compounds and oxo-steroids3 • 1-(3-chlorophenyl)-5-trifluoromethyl-3-hydrobenzimidazol-2-one
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 367-86-2 | |
| 209.1 | |
| HLDFCCHSOZWKAA-UHFFFAOYSA-N | |
| 67778 | |
| C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])F |
| C7H3F4NO2 | |
| MFCD00007059 | |
| 4-fluoro-3-nitrobenzotrifluoride, 1-fluoro-2-nitro-4-trifluoromethyl benzene, benzene, 1-fluoro-2-nitro-4-trifluoromethyl, 4-fluoro-3-nitrotrifluoromethylbenzene, 3-nitro-4-fluorobenzotrifluoride, 3-nitro-alpha,alpha,alpha,4-tetrafluorotoluene, 2-fluoro-5-trifluoromethyl nitrobenzene, 3-nitro-4-fluoro benzotrifluoride, 1-fluoro-2-nitro-4-trifluoromethyl-benzene | |
| 1-fluoro-2-nitro-4-(trifluoromethyl)benzene |
Specifications
| 367-86-2 | |
| 91°C to 92°C (15 mmHg) | |
| C7H3F4NO2 | |
| MFCD00007059 | |
| UN2810 | |
| 4-fluoro-3-nitrobenzotrifluoride, 1-fluoro-2-nitro-4-trifluoromethyl benzene, benzene, 1-fluoro-2-nitro-4-trifluoromethyl, 4-fluoro-3-nitrotrifluoromethylbenzene, 3-nitro-4-fluorobenzotrifluoride, 3-nitro-alpha,alpha,alpha,4-tetrafluorotoluene, 2-fluoro-5-trifluoromethyl nitrobenzene, 3-nitro-4-fluoro benzotrifluoride, 1-fluoro-2-nitro-4-trifluoromethyl-benzene | |
| HLDFCCHSOZWKAA-UHFFFAOYSA-N | |
| 1-fluoro-2-nitro-4-(trifluoromethyl)benzene | |
| 67778 | |
| 97% |
| 1.494 | |
| 90°C (194°F) | |
| 1.462 | |
| 25 g | |
| 1880508 | |
| Insoluble in water. | |
| C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])F | |
| 209.1 | |
| 209.1 | |
| 4-Fluoro-3-nitrobenzotrifluoride |
Safety and Handling
GHS H Statement
H311-H227-H302-H332-H315-H319
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Hazard Class: 3; Packaging Group: III
EINECSNumber : 206-702-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only