missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-3-nitrobenzenesulfonamide 98.0+%, TCI America™
$218.10 - $864.92
Chemical Identifiers
| CAS | 406233-31-6 |
|---|---|
| Molecular Formula | C6H5FN2O4S |
| Molecular Weight (g/mol) | 220.174 |
| MDL Number | MFCD08703185 |
| InChI Key | FAYVDRRKPVJSPE-UHFFFAOYSA-N |
| Synonym | 4-fluoro-3-nitro-benzenesulfonamide, 4-fluoro-3-nitrobenzene-1-sulfonamide, benzenesulfonamide, 4-fluoro-3-nitro, 4-fluoro-3-nitrobenzenesulfonamide, 4-fluoro-3-nitrobenzene sulfonamide, 4-f luoro-3-nitrobenzene sulfonamide, benzenesulfonamide,4-fluoro-3-nitro, 4-fluoranyl-3-nitro-benzenesulfonamide |
| PubChem CID | 16782487 |
| IUPAC Name | 4-fluoro-3-nitrobenzenesulfonamide |
| SMILES | C1=CC(=C(C=C1S(=O)(=O)N)[N+](=O)[O-])F |
Chemical Identifiers
| 406233-31-6 | |
| 220.174 | |
| FAYVDRRKPVJSPE-UHFFFAOYSA-N | |
| 16782487 | |
| C1=CC(=C(C=C1S(=O)(=O)N)[N+](=O)[O-])F |
| C6H5FN2O4S | |
| MFCD08703185 | |
| 4-fluoro-3-nitro-benzenesulfonamide, 4-fluoro-3-nitrobenzene-1-sulfonamide, benzenesulfonamide, 4-fluoro-3-nitro, 4-fluoro-3-nitrobenzenesulfonamide, 4-fluoro-3-nitrobenzene sulfonamide, 4-f luoro-3-nitrobenzene sulfonamide, benzenesulfonamide,4-fluoro-3-nitro, 4-fluoranyl-3-nitro-benzenesulfonamide | |
| 4-fluoro-3-nitrobenzenesulfonamide |
Specifications
| 406233-31-6 | |
| Yellow | |
| MFCD08703185 | |
| 4-fluoro-3-nitro-benzenesulfonamide, 4-fluoro-3-nitrobenzene-1-sulfonamide, benzenesulfonamide, 4-fluoro-3-nitro, 4-fluoro-3-nitrobenzenesulfonamide, 4-fluoro-3-nitrobenzene sulfonamide, 4-f luoro-3-nitrobenzene sulfonamide, benzenesulfonamide,4-fluoro-3-nitro, 4-fluoranyl-3-nitro-benzenesulfonamide | |
| C1=CC(=C(C=C1S(=O)(=O)N)[N+](=O)[O-])F | |
| 220.174 | |
| 220.17 | |
| Crystalline Powder |
| 139°C | |
| C6H5FN2O4S | |
| 1 g | |
| FAYVDRRKPVJSPE-UHFFFAOYSA-N | |
| 4-fluoro-3-nitrobenzenesulfonamide | |
| 16782487 | |
| ≥98.0% (HPLC,N) | |
| 4-Fluoro-3-nitrobenzenesulfonamide |
Safety and Handling
TSCA : No