accessibility menu, dialog, popup

UserName

CAS RN 406233-31-6

IUPAC Name: 4-fluoro-3-nitrobenzene-1-sulfonamide
Synonyms: 4-fluoro-3-nitrobenzenesulfonamide
Molecular Weight (g/mol): 220.17
Molecular Formula: C6H5FN2O4S
InChi Key: FAYVDRRKPVJSPE-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC(=C(F)C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)