missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-nitrobenzenesulfonamide 98.0+%, TCI America™

Catalog No. F11721G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F11721G 1 g
F11725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F11721G Supplier TCI America Supplier No. F11721G

Chemical Identifiers

CAS 406233-31-6
Molecular Formula C6H5FN2O4S
Molecular Weight (g/mol) 220.174
MDL Number MFCD08703185
InChI Key FAYVDRRKPVJSPE-UHFFFAOYSA-N
Synonym 4-fluoro-3-nitro-benzenesulfonamide, 4-fluoro-3-nitrobenzene-1-sulfonamide, benzenesulfonamide, 4-fluoro-3-nitro, 4-fluoro-3-nitrobenzenesulfonamide, 4-fluoro-3-nitrobenzene sulfonamide, 4-f luoro-3-nitrobenzene sulfonamide, benzenesulfonamide,4-fluoro-3-nitro, 4-fluoranyl-3-nitro-benzenesulfonamide
PubChem CID 16782487
IUPAC Name 4-fluoro-3-nitrobenzenesulfonamide
SMILES C1=CC(=C(C=C1S(=O)(=O)N)[N+](=O)[O-])F

Specifications

Melting Point 139°C
Color Yellow
Quantity 1 g
Formula Weight 220.17
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-3-nitrobenzenesulfonamide