missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-nitrobenzaldehyde 95.0+%, TCI America™

Catalog No. F09561G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09561G 1 g
F09565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09561G Supplier TCI America Supplier No. F09561G

Chemical Identifiers

CAS 42564-51-2
Molecular Formula C7H4FNO3
Molecular Weight (g/mol) 169.111
MDL Number MFCD01861388
InChI Key ILKWFRCNNILIJW-UHFFFAOYSA-N
Synonym 3-nitro-4-fluorobenzaldehyde, benzaldehyde, 4-fluoro-3-nitro, 4-fluoro-3-nitro-benzaldehyde, pubchem22572, acmc-1ao2q, 3-nitro-4 fluorobenzaldehyde, ksc497m9r, 2-fluoro-5-formylnitrobenzene, #, 4-fluoranyl-3-nitro-benzaldehyde
PubChem CID 598129
IUPAC Name 4-fluoro-3-nitrobenzaldehyde
SMILES C1=CC(=C(C=C1C=O)[N+](=O)[O-])F

Specifications

Melting Point 45°C
Color Yellow
Quantity 1 g
Formula Weight 169.11
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-3-nitrobenzaldehyde