missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-(methylcarbamoyl)benzeneboronic acid, 96%

CAS: 874219-19-9 | C8H9BFNO3 | 196.97 g/mol

$233.01 - $744.96

Chemical Identifiers

CAS 874219-19-9
Molecular Formula C8H9BFNO3
Molecular Weight (g/mol) 196.97
MDL Number MFCD08235038
InChI Key DVDSMLPVTUSIFM-UHFFFAOYSA-N
Synonym 4-fluoro-3-methylcarbamoyl phenyl boronic acid, 4-fluoro-3-methylcarbamoyl phenylboronic acid, 4-fluoro-3-methylcarbamoyl benzeneboronic acid, acmc-209qkk, n-methyl 5-borono-2-fluorobenzamide, 4-fluoro-3-methylcarbamoyl phenyl boronicacid, 4-fluoro-3-methyl carbamoyl phenylboronic acid, 4-fluoranyl-3-methylcarbamoyl phenyl boronic acid, boronic acid, 4-fluoro-3-methylamino carbonyl phenyl
PubChem CID 44717580
IUPAC Name [4-fluoro-3-(methylcarbamoyl)phenyl]boronic acid
SMILES CNC(=O)C1=C(F)C=CC(=C1)B(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH53101MD
View Documents
Thermo Scientific Chemicals
H53101MD
250 mg
Each for $233.01
Only null left
 
AAH5310103
View Documents
Thermo Scientific Chemicals
H5310103
1 g
Each for $744.96
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

874219-19-9
196.97
DVDSMLPVTUSIFM-UHFFFAOYSA-N
44717580
CNC(=O)C1=C(F)C=CC(=C1)B(O)O
C8H9BFNO3
MFCD08235038
4-fluoro-3-methylcarbamoyl phenyl boronic acid, 4-fluoro-3-methylcarbamoyl phenylboronic acid, 4-fluoro-3-methylcarbamoyl benzeneboronic acid, acmc-209qkk, n-methyl 5-borono-2-fluorobenzamide, 4-fluoro-3-methylcarbamoyl phenyl boronicacid, 4-fluoro-3-methyl carbamoyl phenylboronic acid, 4-fluoranyl-3-methylcarbamoyl phenyl boronic acid, boronic acid, 4-fluoro-3-methylamino carbonyl phenyl
[4-fluoro-3-(methylcarbamoyl)phenyl]boronic acid

Specifications

874219-19-9
C8H9BFNO3
250 mg
DVDSMLPVTUSIFM-UHFFFAOYSA-N
[4-fluoro-3-(methylcarbamoyl)phenyl]boronic acid
44717580
96%
226°C to 228°C
MFCD08235038
4-fluoro-3-methylcarbamoyl phenyl boronic acid, 4-fluoro-3-methylcarbamoyl phenylboronic acid, 4-fluoro-3-methylcarbamoyl benzeneboronic acid, acmc-209qkk, n-methyl 5-borono-2-fluorobenzamide, 4-fluoro-3-methylcarbamoyl phenyl boronicacid, 4-fluoro-3-methyl carbamoyl phenylboronic acid, 4-fluoranyl-3-methylcarbamoyl phenyl boronic acid, boronic acid, 4-fluoro-3-methylamino carbonyl phenyl
CNC(=O)C1=C(F)C=CC(=C1)B(O)O
196.97
196.97
4-Fluoro-3-(methylcarbamoyl)benzeneboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only