accessibility menu, dialog, popup

UserName

CAS RN 874219-19-9

BFOHNCH3HOOH

CAS RN 874219-19-9

IUPAC Name: [4-fluoro-3-(methylcarbamoyl)phenyl]boronic acid
Synonyms: 4-fluoro-3-(methylcarbamoyl)phenylboronic acid
Molecular Weight (g/mol): 196.97
Molecular Formula: C8H9BFNO3
InChi Key: DVDSMLPVTUSIFM-UHFFFAOYSA-N
SMILES: CNC(=O)C1=C(F)C=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results