missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methoxybenzaldehyde, 98%

Catalog No. AAL1967106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1967106 5 g
AAL1967103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1967106 Supplier Thermo Scientific Chemicals Supplier No. L1967106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 128495-46-5 | C8H7FO2 | 154.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 128495-46-5
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00143320
InChI Key NALVGTOMKSKFFV-UHFFFAOYSA-N
Synonym 3-methoxy-4-fluorobenzaldehyde, 4-fluoro-m-anisaldehyde, 4-fluoro-3-methoxy-benzaldehyde, benzaldehyde, 4-fluoro-3-methoxy, pubchem1449, acmc-209bf6, ksc490m2d, 3-methoxy-4-fluoro benzaldehyde, 3-methoxy-4-fluoro-benzaldehyde, nalvgtomkskffv-uhfffaoysa
PubChem CID 2737358
IUPAC Name 4-fluoro-3-methoxybenzaldehyde
SMILES COC1=CC(C=O)=CC=C1F

Specifications

Melting Point 61°C to 63°C
Quantity 5 g
Beilstein 5178063
Sensitivity Air Sensitive
Formula Weight 154.14
Percent Purity 98%
Chemical Name or Material 4-Fluoro-3-methoxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only