missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-3-formylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$61.51 - $61.51
Specifications
| CAS | 374538-01-9 |
|---|---|
| Melting Point | 220°C |
| Color | White-Yellow |
| Molecular Formula | C7H6BFO3 |
| MDL Number | MFCD02093074 |
Specifications
| 374538-01-9 | |
| White-Yellow | |
| MFCD02093074 | |
| B(C1=CC(=C(C=C1)F)C=O)(O)O | |
| 167.93 | |
| 167.93 |
| 220°C | |
| C7H6BFO3 | |
| YABSTJQEBSKPCG-UHFFFAOYSA-N | |
| (4-fluoro-3-formylphenyl)boronic acid | |
| 2778651 | |
| Crystalline Powder |