accessibility menu, dialog, popup

UserName

CAS RN 374538-01-9

HOBOHFO

CAS RN 374538-01-9

IUPAC Name: (4-fluoro-3-formylphenyl)boronic acid
Synonyms: 4-fluoro-3-formylphenylboronic acid
Molecular Weight (g/mol): 167.93
Molecular Formula: C7H6BFO3
InChi Key: YABSTJQEBSKPCG-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(F)C(C=O)=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results