missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-(trifluoromethyl)benzyl bromide, 98%, Thermo Scientific™

Catalog No. AC434220010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434220010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC434220010 Supplier Thermo Scientific Chemicals Supplier No. 434220010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 206860-48-2
Molecular Formula C8H5BrF4
Molecular Weight (g/mol) 257.03
InChI Key JMNOONULDANZRZ-UHFFFAOYSA-N
Synonym 4-fluoro-2-trifluoromethyl benzyl bromide, 1-bromomethyl-4-fluoro-2-trifluoromethyl benzene, 4-fluoro-2-trifluoromethyl benzylbromide, benzene, 1-bromomethyl-4-fluoro-2-trifluoromethyl, alpha-bromo-4-fluoro-2-trifluoromethyl toluene, 1-bromomethyl-4-fluoro-2-trifluoromethyl-benzene, pubchem4642, acmc-20ai28, 4-fluoro-2-trifluoromethylbenzyl bromide, 4-fluoro-2-trifluoromethyl-benzyl bromide
PubChem CID 2737576
IUPAC Name 1-(bromomethyl)-4-fluoro-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1F)C(F)(F)F)CBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 141°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 257.03
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)benzyl bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER

RUO – Research Use Only