CAS RN 206860-48-2
IUPAC Name:
1-(bromomethyl)-2-fluoro-4-(trifluoromethyl)benzene
Synonyms:
1-(bromomethyl)-2-fluoro-4-(trifluoromethyl)benzene
Molecular Weight (g/mol):
257.03
Molecular Formula:
C8H5BrF4
InChi Key:
MQUCRCQNRDHRPL-UHFFFAOYSA-N
SMILES:
FC1=CC(=CC=C1CBr)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
4-Fluoro-2-(trifluoromethyl)benzyl bromide, 98%, Thermo Scientific™
4-Fluoro-2-(trifluoromethyl)benzyl bromide, 98%, Thermo Scientific™
CAS: 206860-48-2 Molecular Formula: C8H5BrF4 Molecular Weight (g/mol): 257.03 InChI Key: JMNOONULDANZRZ-UHFFFAOYSA-N Synonym: 4-fluoro-2-trifluoromethyl benzyl bromide,1-bromomethyl-4-fluoro-2-trifluoromethyl benzene,4-fluoro-2-trifluoromethyl benzylbromide,benzene, 1-bromomethyl-4-fluoro-2-trifluoromethyl,alpha-bromo-4-fluoro-2-trifluoromethyl toluene,1-bromomethyl-4-fluoro-2-trifluoromethyl-benzene,pubchem4642,acmc-20ai28,4-fluoro-2-trifluoromethylbenzyl bromide,4-fluoro-2-trifluoromethyl-benzyl bromide PubChem CID: 2737576 IUPAC Name: 1-(bromomethyl)-4-fluoro-2-(trifluoromethyl)benzene SMILES: C1=CC(=C(C=C1F)C(F)(F)F)CBr