missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-nitrobenzonitrile, 97%

Catalog No. p-7031509
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
10 g
Catalog No. Quantity
AAH3261003 1 g
AAH3261009 10 g
2 options

Catalog No. AAH3261003

Supplier: Thermo Scientific Chemicals H3261003

Only null left

CAS: 80517-21-1 | C7H3FN2O2 | 166.111 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80517-21-1
Molecular Formula C7H3FN2O2
Molecular Weight (g/mol) 166.111
MDL Number MFCD00277447
InChI Key MNEAKKQYFSYZEU-UHFFFAOYSA-N
Synonym 2-cyano-5-fluoronitrobenzene, benzonitrile, 4-fluoro-2-nitro, buttpark 100\07-73, pubchem4795, acmc-209pjl, 4-fluoro-2-nitrobenzonitrle, ksc497q7l, 4-fluoro-2-nitro-benzonitrile, benzonitrile,4-fluoro-2-nitro, 4-fluoro-2-nitrobenzenecarbonitrile
PubChem CID 2774652
IUPAC Name 4-fluoro-2-nitrobenzonitrile
SMILES C1=CC(=C(C=C1F)[N+](=O)[O-])C#N

Specifications

Melting Point 69°C to 73°C
Quantity 1 g
UN Number UN3439
Formula Weight 166.11
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-nitrobenzonitrile
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only