accessibility menu, dialog, popup

UserName

CAS RN 80517-21-1

IUPAC Name: 4-fluoro-2-nitrobenzonitrile
Synonyms: 4-fluoro-2-nitrobenzonitrile
Molecular Weight (g/mol): 166.11
Molecular Formula: C7H3FN2O2
InChi Key: MNEAKKQYFSYZEU-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(C=CC(F)=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results