missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-nitrobenzaldehyde, 98%

Catalog No. p-7041399
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6197103 1 g
AAH6197106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6197103 Supplier Thermo Scientific Chemicals Supplier No. H6197103

CAS: 2923-96-8 | C7H4FNO3 | 169.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2923-96-8
Molecular Formula C7H4FNO3
Molecular Weight (g/mol) 169.11
MDL Number MFCD01666439
InChI Key ORCGMGUNVGVHDN-UHFFFAOYSA-N
Synonym 4-fluoro-2-nitro-benzaldehyde, benzaldehyde, 4-fluoro-2-nitro, pubchem15643, acmc-1chrz, wln: wnr cf fvh, 2-nitro-4-fluorobenzaldehyde, ksc494k6p, 4-fluoro-2-nitro benzaldehyde
PubChem CID 76226
IUPAC Name 4-fluoro-2-nitrobenzaldehyde
SMILES [O-][N+](=O)C1=CC(F)=CC=C1C=O

Specifications

Quantity 1 g
Formula Weight 169.11
Percent Purity 98%
Chemical Name or Material 4-Fluoro-2-nitrobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-884-3
RTECSNumber CU6250000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only