missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-nitroanisole 98.0+%, TCI America™

Catalog No. p-7129785
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06155G 5 g
F061525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06155G Supplier TCI America Supplier No. F06155G

Chemical Identifiers

CAS 445-83-0
Molecular Formula C7H6FNO3
Molecular Weight (g/mol) 171.13
MDL Number MFCD00013375
InChI Key FWLPYISRFBKEKV-UHFFFAOYSA-N
Synonym 4-fluoro-2-nitroanisole, 4-fluoro-2-nitroanisol, benzene, 4-fluoro-1-methoxy-2-nitro, 4-fluoro-1-methoxy-2-nitro-benzene, 5-fluoro-2-methoxynitrobenzene, 4-fluor-1-methoxy-2-nitrobenzol, pubchem1966, acmc-1amcp, 4-fluoro-2-nitro-anisole, ksc235o8j
PubChem CID 565656
IUPAC Name 4-fluoro-1-methoxy-2-nitrobenzene
SMILES COC1=C(C=C(F)C=C1)[N+]([O-])=O

Specifications

Melting Point 63°C
Color Yellow
Quantity 5 g
Formula Weight 171.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-2-nitroanisole