accessibility menu, dialog, popup

UserName

H3COFNOO

CAS RN 445-83-0

IUPAC Name: 4-fluoro-1-methoxy-2-nitrobenzene
Synonyms: 4-fluoro-1-methoxy-2-nitrobenzene
Molecular Weight (g/mol): 171.13
Molecular Formula: C7H6FNO3
InChi Key: FWLPYISRFBKEKV-UHFFFAOYSA-N
SMILES: COC1=C(C=C(F)C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)