missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific™

Catalog No. AC431840500
Change view
Click to view available options
Quantity:
50mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC431840500 50mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC431840500 Supplier Thermo Scientific Chemicals Supplier No. 431840500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 30897-90-6 , 109-99-9
Molecular Formula C7H6BrFMg
Molecular Weight (g/mol) 213.33
MDL Number MFCD01311469
InChI Key OCGINFOPBMDLBD-UHFFFAOYSA-M
Synonym 4-fluoro-2-methylphenylmagnesium bromide, bromo 4-fluoro-2-methylphenyl magnesium, swsqupddggyhlr-uhfffaoysa-m, 4-fluoro-2-methylphenyl magnesium bromide, 4-fluoro-2-methyl-phenyl magnesium bromide, 4-fluoro-2-methylphenylmagnesium bromide, 0.50 m in 2-methf, 4-fluoro-2-methylphenylmagnesium bromide 0.5 m in tetrahydrofuran, 4-fluoro-2-methylphenylmagnesium bromide solution, 0.5 m in thf, grignard reagent
PubChem CID 2778803
IUPAC Name magnesium;1-fluoro-3-methylbenzene-4-ide;bromide
SMILES CC1=CC(=CC=[C-]1)F.[Mg+2].[Br-]

Specifications

CAS Min % 8.91, 86.64
CAS Max % 13.36, 91.09
Density 0.9580g/mL
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Linear Formula F(CH3)C6H3MgBr
Quantity 50mL
Specific Gravity 0.958
Solubility Information Solubility in water: reacts
Formula Weight 213.33
Chemical Name or Material 4-Fluoro-2-methylphenylmagnesium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br/
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only