accessibility menu, dialog, popup

UserName

CAS RN 30897-90-6

IUPAC Name: bromo(4-fluoro-2-methylphenyl)magnesium
Synonyms: bromo(4-fluoro-2-methylphenyl)magnesium
Molecular Weight (g/mol): 213.33
Molecular Formula: C7H6BrFMg
InChi Key: SWSQUPDDGGYHLR-UHFFFAOYSA-M
SMILES: CC1=CC(F)=CC=C1[Mg]Br
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results