missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylaniline 98.0+%, TCI America™

Catalog No. p-7129661
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03985G 5 g
F039825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03985G Supplier TCI America Supplier No. F03985G
Only null left

Chemical Identifiers

CAS 452-71-1
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.15
MDL Number MFCD00007832
InChI Key KMHLGVTVACLEJE-UHFFFAOYSA-N
Synonym 2-amino-5-fluorotoluene, benzenamine, 4-fluoro-2-methyl, 4-fluoro-2-methylbenzenamine, 6-amino-3-fluorotoluene, o-toluidine, 4-fluoro, 2-methyl-4-fluoroaniline, 4-fluorotoluidine, 4-fluoro-o-toluidine, 3-fluoro-6-aminotoluene
PubChem CID 67982
IUPAC Name 4-fluoro-2-methylaniline
SMILES CC1=CC(F)=CC=C1N

Specifications

Color Yellow
Boiling Point 102°C
Quantity 5 g
Formula Weight 125.15
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Fluoro-2-methylaniline
RTECSNumber CY0500000
TSCA Yes