missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylaniline, 97%

Catalog No. p-7081578
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
Catalog No. Quantity
AAA1160914 25 g
1 options

Catalog No. AAA1160914

Supplier: Thermo Scientific Chemicals A1160914

Only null left

CAS: 452-71-1 | C7H8FN | 125.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 452-71-1
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.15
MDL Number MFCD00007832
InChI Key KMHLGVTVACLEJE-UHFFFAOYSA-N
Synonym 2-amino-5-fluorotoluene, benzenamine, 4-fluoro-2-methyl, 4-fluoro-2-methylbenzenamine, 6-amino-3-fluorotoluene, o-toluidine, 4-fluoro, 2-methyl-4-fluoroaniline, 4-fluorotoluidine, 4-fluoro-o-toluidine, 3-fluoro-6-aminotoluene
PubChem CID 67982
IUPAC Name 4-fluoro-2-methylaniline
SMILES CC1=CC(F)=CC=C1N

Specifications

Density 1.124
Boiling Point 90°C to 92°C (16 mmHg)
Flash Point 87°C (188°F)
Odor Characteristic
Refractive Index 1.537
Quantity 25 g
UN Number UN2810
Beilstein 1931798
Formula Weight 125.15
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-methylaniline
Show More Show Less
Hazard Category H227-H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H311-H373-H227-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 207-208-2
RTECSNumber CY0500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only