missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-iodobenzoic acid, 97%

Catalog No. p-7037008
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5553103 1 g
AAH5553106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5553103 Supplier Thermo Scientific Chemicals Supplier No. H5553103

CAS: 56096-89-0 | C7H4FIO2 | 266.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56096-89-0
Molecular Formula C7H4FIO2
Molecular Weight (g/mol) 266.01
MDL Number MFCD04108278
InChI Key DUKFTVLJAXWGPI-UHFFFAOYSA-N
Synonym 4-fluoro-2-iodo-benzoic acid, 2-iodo-4-fluorobenzoic acid, 4-fluoro-2-iodobenzoicacid, benzoic acid, 4-fluoro-2-iodo, benzoic acid, 4-fluoroiodo, pubchem1389, acmc-1atr7, ksc494a9j, benzoic acid,4-fluoro-2-iodo, buttpark 83\07-33
PubChem CID 12520164
IUPAC Name 4-fluoro-2-iodobenzoic acid
SMILES C1=CC(=C(C=C1F)I)C(=O)O

Specifications

Melting Point 143°C to 147°C
Quantity 1 g
UN Number UN2811
Sensitivity Light sensitive
Formula Weight 266.01
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-iodobenzoic acid
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only