CAS RN 56096-89-0
IUPAC Name:
4-fluoro-2-iodobenzoic acid
Synonyms:
4-fluoro-2-iodobenzoic acid
Molecular Weight (g/mol):
266.01
Molecular Formula:
C7H4FIO2
InChi Key:
DUKFTVLJAXWGPI-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C(I)C=C(F)C=C1
1
–
1
of
1
results
1
Promotions Available
4-Fluoro-2-iodobenzoic acid, 97%
4-Fluoro-2-iodobenzoic acid, 97%
CAS: 56096-89-0 Molecular Formula: C7H4FIO2 Molecular Weight (g/mol): 266.01 MDL Number: MFCD04108278 InChI Key: DUKFTVLJAXWGPI-UHFFFAOYSA-N Synonym: 4-fluoro-2-iodo-benzoic acid,2-iodo-4-fluorobenzoic acid,4-fluoro-2-iodobenzoicacid,benzoic acid, 4-fluoro-2-iodo,benzoic acid, 4-fluoroiodo,pubchem1389,acmc-1atr7,ksc494a9j,benzoic acid,4-fluoro-2-iodo,buttpark 83\07-33 PubChem CID: 12520164 IUPAC Name: 4-fluoro-2-iodobenzoic acid SMILES: C1=CC(=C(C=C1F)I)C(=O)O