missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-2-iodoaniline 98.0+%, TCI America™
$89.48 - $301.98
Chemical Identifiers
| CAS | 61272-76-2 |
|---|---|
| Molecular Formula | C6H5FIN |
| Molecular Weight (g/mol) | 237.02 |
| MDL Number | MFCD00042584 |
| InChI Key | SETOTRGVPANENO-UHFFFAOYSA-N |
| Synonym | 2-iodo-4-fluoroaniline, 4-fluoro-2-indoaniline, 4-fluoro-2-iodoanline, 4-fluoro-2-iodo-aniline, 4-fluoro-2-iodophenylamine, 4-fluoro-2-iodo-phenylamine, benzenamine, 4-fluoro-2-iodo, zlchem 368, pubchem3413, 2-iodo-4-fluoro-aniline |
| PubChem CID | 2778283 |
| IUPAC Name | 4-fluoro-2-iodoaniline |
| SMILES | NC1=C(I)C=C(F)C=C1 |
Chemical Identifiers
| 61272-76-2 | |
| 237.02 | |
| SETOTRGVPANENO-UHFFFAOYSA-N | |
| 2778283 | |
| NC1=C(I)C=C(F)C=C1 |
| C6H5FIN | |
| MFCD00042584 | |
| 2-iodo-4-fluoroaniline, 4-fluoro-2-indoaniline, 4-fluoro-2-iodoanline, 4-fluoro-2-iodo-aniline, 4-fluoro-2-iodophenylamine, 4-fluoro-2-iodo-phenylamine, benzenamine, 4-fluoro-2-iodo, zlchem 368, pubchem3413, 2-iodo-4-fluoro-aniline | |
| 4-fluoro-2-iodoaniline |
Specifications
| 61272-76-2 | |
| MFCD00042584 | |
| 2-iodo-4-fluoroaniline, 4-fluoro-2-indoaniline, 4-fluoro-2-iodoanline, 4-fluoro-2-iodo-aniline, 4-fluoro-2-iodophenylamine, 4-fluoro-2-iodo-phenylamine, benzenamine, 4-fluoro-2-iodo, zlchem 368, pubchem3413, 2-iodo-4-fluoro-aniline | |
| NC1=C(I)C=C(F)C=C1 | |
| 237.02 | |
| 237.02 | |
| Liquid |
| C6H5FIN | |
| 5 g | |
| SETOTRGVPANENO-UHFFFAOYSA-N | |
| 4-fluoro-2-iodoaniline | |
| 2778283 | |
| ≥98.0% (GC,T) | |
| 4-Fluoro-2-iodoaniline |
Safety and Handling
TSCA : No