missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-iodo-1-nitrobenzene 98.0+%, TCI America™

Catalog No. p-7130091
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F11781G 1 g
F11785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F11781G Supplier TCI America Supplier No. F11781G

Chemical Identifiers

CAS 41860-64-4
Molecular Formula C6H3FINO2
Molecular Weight (g/mol) 267.00
MDL Number MFCD16658618
InChI Key QKROTSNREAHREK-UHFFFAOYSA-N
PubChem CID 19361948
IUPAC Name 4-fluoro-2-iodo-1-nitrobenzene
SMILES [O-][N+](=O)C1=C(I)C=C(F)C=C1

Specifications

Melting Point 30°C
Boiling Point 148°C
Quantity 1 g
Formula Weight 267.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Fluoro-2-iodo-1-nitrobenzene
TSCA No
Recommended Storage Refrigerator