accessibility menu, dialog, popup

UserName

FINOO

CAS RN 41860-64-4

IUPAC Name: 4-fluoro-2-iodo-1-nitrobenzene
Synonyms: 4-fluoro-2-iodo-1-nitrobenzene
Molecular Weight (g/mol): 267
Molecular Formula: C6H3FINO2
InChi Key: QKROTSNREAHREK-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(I)C=C(F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results