missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-hydroxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Chemical Identifiers

CAS 1038828-32-8
Molecular Formula C12H16BFO3
Molecular Weight (g/mol) 238.07
MDL Number MFCD16994248
InChI Key PSQUBOBJDBVJIN-UHFFFAOYSA-N
Synonym 5-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-fluoro-2-hydroxybenzeneboronic acid pinacol ester, 4-fluoro-2-hydroxyphenylboronic acid pinacol ester, phenol, 5-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 72219414
IUPAC Name 5-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
SMILES CC1(C)OB(OC1(C)C)C1=C(O)C=C(F)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAH62252MD
View Documents Promotion Details
N/A 250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6225203
View Documents Promotion Details
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1038828-32-8
238.07
PSQUBOBJDBVJIN-UHFFFAOYSA-N
72219414
CC1(C)OB(OC1(C)C)C1=C(O)C=C(F)C=C1
C12H16BFO3
MFCD16994248
5-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-fluoro-2-hydroxybenzeneboronic acid pinacol ester, 4-fluoro-2-hydroxyphenylboronic acid pinacol ester, phenol, 5-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
5-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

Specifications

1038828-32-8
MFCD16994248
PSQUBOBJDBVJIN-UHFFFAOYSA-N
5-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
72219414
96%
C12H16BFO3
5-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-fluoro-2-hydroxybenzeneboronic acid pinacol ester, 4-fluoro-2-hydroxyphenylboronic acid pinacol ester, phenol, 5-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
CC1(C)OB(OC1(C)C)C1=C(O)C=C(F)C=C1
238.07
238.06
4-Fluoro-2-hydroxybenzeneboronic acid pinacol ester
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents