missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-hydroxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H6225203

Quantity 1g

 View more versions of this product

Catalog No. AAH6225203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1038828-32-8
238.07
PSQUBOBJDBVJIN-UHFFFAOYSA-N
72219414
CC1(C)OB(OC1(C)C)C1=C(O)C=C(F)C=C1
C12H16BFO3
MFCD16994248
5-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-fluoro-2-hydroxybenzeneboronic acid pinacol ester, 4-fluoro-2-hydroxyphenylboronic acid pinacol ester, phenol, 5-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
5-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

Specifications

4-Fluoro-2-hydroxybenzeneboronic acid pinacol ester
C12H16BFO3
1g
PSQUBOBJDBVJIN-UHFFFAOYSA-N
5-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
72219414
96%
1038828-32-8
MFCD16994248
5-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-fluoro-2-hydroxybenzeneboronic acid pinacol ester, 4-fluoro-2-hydroxyphenylboronic acid pinacol ester, phenol, 5-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
CC1(C)OB(OC1(C)C)C1=C(O)C=C(F)C=C1
238.07
238.06
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions