missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2,6-dimethylaniline, 97%

Catalog No. p-7027904
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2496503 1 g
AAB2496506 5 g
AAB2496514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2496503 Supplier Thermo Scientific Chemicals Supplier No. B2496503

CAS: 392-70-1 | C8H10FN | 139.173 g/mol

4-Fluoro-2,6-dimethylaniline is used as an intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-2,6-dimethylaniline is used as an intermediate.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 392-70-1
Molecular Formula C8H10FN
Molecular Weight (g/mol) 139.173
MDL Number MFCD03792669
InChI Key MPNDLCBMLBACDD-UHFFFAOYSA-N
Synonym 4-fluoro-2,6-dimethylphenylamine, 2,6-dimethyl-4-fluoroaniline, acmc-20amil, pubchem4426, 4-fluoro-2,6-dimethyl-aniline, 4-fluoranyl-2,6-dimethyl-aniline, benzenamine,4-fluoro-2,6-dimethyl
PubChem CID 1403908
IUPAC Name 4-fluoro-2,6-dimethylaniline
SMILES CC1=CC(=CC(=C1N)C)F

Specifications

Quantity 1 g
Refractive Index 1.5345
Formula Weight 139.17
Percent Purity 98%
Chemical Name or Material 4-Fluoro-2,6-dimethylaniline
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only