missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-1,2-phenylenediamine 98.0+%, TCI America™

Catalog No. D37265G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D37265G Supplier TCI America Supplier No. D37265G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 367-31-7
Molecular Formula C6H7FN2
Molecular Weight (g/mol) 126.134
MDL Number MFCD00042228
InChI Key KWEWNOOZQVJONF-UHFFFAOYSA-N
Synonym 1,2-diamino-4-fluorobenzene, 4-fluoro-1,2-phenylenediamine, 3,4-diaminofluorobenzene, 4-fluoro-o-phenylenediamine, 2-amino-4-fluoroaniline, 4-fluoro-1,2-diaminobenzene, 3,4-diamino-1-fluorobenzene, 5-fluoro-1,2-diaminobenzene, 4-fluoro-1,2-benzenediamine, 1,2-benzenediamine, 4-fluoro
PubChem CID 164584
IUPAC Name 4-fluorobenzene-1,2-diamine
SMILES C1=CC(=C(C=C1F)N)N

Specifications

Melting Point 94°C
Quantity 5 g
Formula Weight 126.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-1,2-phenylenediamine