missing translation for 'onlineSavingsMsg'
Learn More

4-(Ethylthio)benzaldehyde, 97+%

Catalog No. p-7047109
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1762403 1 g
AAL1762406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1762403 Supplier Thermo Scientific Chemicals Supplier No. L1762403
Only null left

CAS: 84211-94-9 | C9H10OS | 166.238 g/mol

It is an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical ingredient.

Solubility
Difficult to mix in water.

Notes
Air sensitive. Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 84211-94-9
Molecular Formula C9H10OS
Molecular Weight (g/mol) 166.238
MDL Number MFCD01318145
InChI Key LPRKZLUVZOFDDT-UHFFFAOYSA-N
Synonym 4-ethylthio benzaldehyde, 4-ethylsulfanyl benzaldehyde, 4-ethylthiobenzaldehyde, 4-ethylsulfanyl-benzaldehyde, benzaldehyde,4-ethylthio
PubChem CID 7006654
IUPAC Name 4-ethylsulfanylbenzaldehyde
SMILES CCSC1=CC=C(C=C1)C=O

Specifications

Boiling Point 151°C to 153°C (15 mmHg)
Refractive Index 1.624
Quantity 1 g
Beilstein 1863857
Sensitivity Air Sensitive
Solubility Information Difficult to mix in water.
Formula Weight 166.24
Percent Purity 98%
Chemical Name or Material 4-(Ethylthio)benzaldehyde
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only