accessibility menu, dialog, popup

UserName

CAS RN 84211-94-9

IUPAC Name: 4-(ethylsulfanyl)benzaldehyde
Synonyms: 4-(ethylsulfanyl)benzaldehyde
Molecular Weight (g/mol): 166.24
Molecular Formula: C9H10OS
InChi Key: LPRKZLUVZOFDDT-UHFFFAOYSA-N
SMILES: CCSC1=CC=C(C=O)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results