Learn More
4-Ethylresorcinol, 98%
CAS: 2896-60-8 | C8H10O2 | 138.166 g/mol
Supplier: Thermo Scientific Chemicals B2157522
Description
It is employed as an active pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as an active pharmaceutical intermediate.
Solubility
Solubility in methanol; almost transparency. Soluble in alcohol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| 4-Ethylresorcinol | |
| 95°C to 98°C | |
| MFCD00002283 | |
| 1238231 | |
| VGMJYYDKPUPTID-UHFFFAOYSA-N | |
| 4-ethylbenzene-1,3-diol | |
| 17927 | |
| 98% |
| 2896-60-8 | |
| C8H10O2 | |
| 100 g | |
| 4-ethylresorcinol, 1,3-benzenediol, 4-ethyl, 6-ethylresorcinol, resorcinol, 4-ethyl, 1,3-dihydroxy-4-ethylbenzene, 4-ethyl resorcinol, 2,4-dihydroxy-1-ethylbenzene, unii-8ye81t06d5, 4-ethyl-benzene-1,3-diol, 4-ethyl-resorcinol | |
| CCC1=C(C=C(C=C1)O)O | |
| 138.166 | |
| 138.17 |
Chemical Identifiers
| 2896-60-8 | |
| 138.166 | |
| VGMJYYDKPUPTID-UHFFFAOYSA-N | |
| 17927 | |
| CCC1=C(C=C(C=C1)O)O |
| C8H10O2 | |
| MFCD00002283 | |
| 4-ethylresorcinol, 1,3-benzenediol, 4-ethyl, 6-ethylresorcinol, resorcinol, 4-ethyl, 1,3-dihydroxy-4-ethylbenzene, 4-ethyl resorcinol, 2,4-dihydroxy-1-ethylbenzene, unii-8ye81t06d5, 4-ethyl-benzene-1,3-diol, 4-ethyl-resorcinol | |
| 4-ethylbenzene-1,3-diol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 220-777-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only