missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylresorcinol, 98%

Catalog No. p-7025866
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2157506 5 g
AAB2157514 25 g
AAB2157522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2157506 Supplier Thermo Scientific Chemicals Supplier No. B2157506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2896-60-8 | C8H10O2 | 138.166 g/mol

It is employed as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an active pharmaceutical intermediate.

Solubility
Solubility in methanol; almost transparency. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 2896-60-8
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00002283
InChI Key VGMJYYDKPUPTID-UHFFFAOYSA-N
Synonym 4-ethylresorcinol, 1,3-benzenediol, 4-ethyl, 6-ethylresorcinol, resorcinol, 4-ethyl, 1,3-dihydroxy-4-ethylbenzene, 4-ethyl resorcinol, 2,4-dihydroxy-1-ethylbenzene, unii-8ye81t06d5, 4-ethyl-benzene-1,3-diol, 4-ethyl-resorcinol
PubChem CID 17927
IUPAC Name 4-ethylbenzene-1,3-diol
SMILES CCC1=C(C=C(C=C1)O)O

Specifications

Melting Point 95°C to 98°C
Quantity 5 g
Beilstein 1238231
Formula Weight 138.17
Percent Purity 98%
Chemical Name or Material 4-Ethylresorcinol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-777-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only