missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethylphenyl Acetate 96.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 226°C |
| Quantity | 25 g |
| Formula Weight | 164.20 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 4-Ethylphenyl Acetate |
Safety and Handling
| TSCA | Yes |