CAS RN 3245-23-6
CAS RN 3245-23-6
IUPAC Name:
4-ethylphenyl acetate
Synonyms:
4-ethylphenyl acetate
Molecular Weight (g/mol):
164.2
Molecular Formula:
C10H12O2
InChi Key:
ANMYMLIUCWWISO-UHFFFAOYSA-N
SMILES:
CCC1=CC=C(OC(C)=O)C=C1
1
–
1
of
1
results
1
4-Ethylphenyl Acetate 96.0+%, TCI America™
CAS: 3245-23-6 Molecular Formula: C10H12O2 Molecular Weight (g/mol): 164.204 MDL Number: MFCD00026970 InChI Key: ANMYMLIUCWWISO-UHFFFAOYSA-N Synonym: Acetic Acid 4-Ethylphenyl Ester PubChem CID: 76731 IUPAC Name: (4-ethylphenyl) acetate SMILES: CCC1=CC=C(C=C1)OC(=O)C