missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylphenol 97.0+%, TCI America™

Catalog No. p-7128868
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E015925G 25 g
E0159500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E015925G Supplier TCI America Supplier No. E015925G
Only null left

Chemical Identifiers

CAS 123-07-9
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00002393
InChI Key HXDOZKJGKXYMEW-UHFFFAOYSA-N
Synonym p-ethylphenol, phenol, 4-ethyl, 1-ethyl-4-hydroxybenzene, phenol, p-ethyl, 1-hydroxy-4-ethylbenzene, para-ethylphenol, 4-hydroxyphenylethane, 4-ethyl-phenol, 4-hydroxyethylbenzene, paraethylphenol
PubChem CID 31242
ChEBI CHEBI:49584
IUPAC Name 4-ethylphenol
SMILES CCC1=CC=C(C=C1)O

Specifications

Color White-Yellow
Boiling Point 219°C
Quantity 25 g
UN Number 2430
Formula Weight 122.17
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Ethylphenol
EINECSNumber (3)-0500&(4)-0057
RTECSNumber SL4040000
TSCA Yes