accessibility menu, dialog, popup

UserName

CAS RN 123-07-9

IUPAC Name: 4-ethylphenol
Synonyms: ethylphenol paraethylphenol P-ethylphenol
Molecular Weight (g/mol): 122.17
Molecular Formula: C8H10O
InChi Key: HXDOZKJGKXYMEW-UHFFFAOYSA-N
SMILES: CCC1=CC=C(O)C=C1
Boiling Point 
  • (1)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)