missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylbenzyl Chloride 98.0+%, TCI America™

Supplier:  TCI America E073825G

 View more versions of this product

Catalog No. E073825G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Ethylbenzyl Chloride
Yellow
C9H11Cl
25 g
4-ethylbenzyl chloride, 1-chloromethyl-4-ethylbenzene, p-ethylbenzylchloride, p-ethylbenzyl chloride, benzene, 1-chloromethyl-4-ethyl, ethyxlbenzylchloride, 4-ethylbenzylchloride, 1-chloromethyl-4-ethyl-benzene, 4-chloromethyl-1-ethylbenzene, 4-ethylbenzyl chloride contains ca 2-form
CCC1=CC=C(CCl)C=C1
154.64
154.64
Liquid
1467-05-6
111°C
MFCD00039357
3265
DUBCVXSYZVTCOC-UHFFFAOYSA-N
1-(chloromethyl)-4-ethylbenzene
73843
≥98.0% (GC)

Chemical Identifiers

1467-05-6
154.64
DUBCVXSYZVTCOC-UHFFFAOYSA-N
73843
CCC1=CC=C(CCl)C=C1
C9H11Cl
MFCD00039357
4-ethylbenzyl chloride, 1-chloromethyl-4-ethylbenzene, p-ethylbenzylchloride, p-ethylbenzyl chloride, benzene, 1-chloromethyl-4-ethyl, ethyxlbenzylchloride, 4-ethylbenzylchloride, 1-chloromethyl-4-ethyl-benzene, 4-chloromethyl-1-ethylbenzene, 4-ethylbenzyl chloride contains ca 2-form
1-(chloromethyl)-4-ethylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions