missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylbenzyl Chloride 98.0+%, TCI America™

Catalog No. p-7129122
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07385G 5 g
E073825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07385G Supplier TCI America Supplier No. E07385G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1467-05-6
Molecular Formula C9H11Cl
Molecular Weight (g/mol) 154.64
MDL Number MFCD00039357
InChI Key DUBCVXSYZVTCOC-UHFFFAOYSA-N
Synonym 4-ethylbenzyl chloride, 1-chloromethyl-4-ethylbenzene, p-ethylbenzylchloride, p-ethylbenzyl chloride, benzene, 1-chloromethyl-4-ethyl, ethyxlbenzylchloride, 4-ethylbenzylchloride, 1-chloromethyl-4-ethyl-benzene, 4-chloromethyl-1-ethylbenzene, 4-ethylbenzyl chloride contains ca 2-form
PubChem CID 73843
IUPAC Name 1-(chloromethyl)-4-ethylbenzene
SMILES CCC1=CC=C(CCl)C=C1

Specifications

Color Yellow
Boiling Point 111°C
Quantity 5 g
UN Number 3265
Formula Weight 154.64
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Ethylbenzyl Chloride