missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Ethylaminomethyl)pyridine 97.0+%, TCI America™
Supplier: TCI America E0283250G
Specifications
| 4-(Ethylaminomethyl)pyridine | |
| Yellow | |
| C8H12N2 | |
| 250 g | |
| ZBAMQLFFVBPAOX-UHFFFAOYSA-N | |
| N-(pyridin-4-ylmethyl)ethanamine | |
| 96681 | |
| ≥97.0% (GC,T) |
| 33403-97-3 | |
| 105°C | |
| MFCD00023632 | |
| 4-ethylaminomethyl pyridine, n-pyridin-4-ylmethyl ethanamine, n-4-pyridylmethyl ethylamine, 4-pyridinemethanamine, n-ethyl, unii-eye32x3w6x, n-ethyl-4-pyridinemethanamine, ethyl pyridin-4-ylmethyl amine, 4-ethylamino methyl pyridine, n-ethyl-4-picolylamine, eye32x3w6x | |
| CCNCC1=CC=NC=C1 | |
| 136.198 | |
| 136.20 | |
| Liquid |
Chemical Identifiers
| 33403-97-3 | |
| 136.198 | |
| ZBAMQLFFVBPAOX-UHFFFAOYSA-N | |
| 96681 | |
| CCNCC1=CC=NC=C1 |
| C8H12N2 | |
| MFCD00023632 | |
| 4-ethylaminomethyl pyridine, n-pyridin-4-ylmethyl ethanamine, n-4-pyridylmethyl ethylamine, 4-pyridinemethanamine, n-ethyl, unii-eye32x3w6x, n-ethyl-4-pyridinemethanamine, ethyl pyridin-4-ylmethyl amine, 4-ethylamino methyl pyridine, n-ethyl-4-picolylamine, eye32x3w6x | |
| N-(pyridin-4-ylmethyl)ethanamine |
Safety and Handling
EINECSNumber : (5)-3700&(9)-0210
TSCA : No