missing translation for 'onlineSavingsMsg'
Learn More

4-(Ethylaminomethyl)pyridine 97.0+%, TCI America™

Catalog No. p-7128917
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E028325G 25 g
E0283250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E028325G Supplier TCI America Supplier No. E028325G
Only null left

Chemical Identifiers

CAS 33403-97-3
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00023632
InChI Key ZBAMQLFFVBPAOX-UHFFFAOYSA-N
Synonym 4-ethylaminomethyl pyridine, n-pyridin-4-ylmethyl ethanamine, n-4-pyridylmethyl ethylamine, 4-pyridinemethanamine, n-ethyl, unii-eye32x3w6x, n-ethyl-4-pyridinemethanamine, ethyl pyridin-4-ylmethyl amine, 4-ethylamino methyl pyridine, n-ethyl-4-picolylamine, eye32x3w6x
PubChem CID 96681
IUPAC Name N-(pyridin-4-ylmethyl)ethanamine
SMILES CCNCC1=CC=NC=C1

Specifications

Color Yellow
Boiling Point 105°C
Quantity 25 g
Formula Weight 136.20
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-(Ethylaminomethyl)pyridine
EINECSNumber (5)-3700&(9)-0210
TSCA No