accessibility menu, dialog, popup

UserName

CAS RN 33403-97-3

IUPAC Name: ethyl[(pyridin-4-yl)methyl]amine
Synonyms: ethyl(pyridin-4-ylmethyl)amine
Molecular Weight (g/mol): 136.2
Molecular Formula: C8H12N2
InChi Key: ZBAMQLFFVBPAOX-UHFFFAOYSA-N
SMILES: CCNCC1=CC=NC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)