missing translation for 'onlineSavingsMsg'
Learn More

4-Ethyl-m-xylene 95.0+%, TCI America™

Catalog No. E02411ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E02411ML 1 mL
E02415ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E02411ML Supplier TCI America Supplier No. E02411ML

Chemical Identifiers

CAS 874-41-9
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00039905
InChI Key MEMBJMDZWKVOTB-UHFFFAOYSA-N
Synonym 4-ethyl-m-xylene, 1,3-dimethyl-4-ethylbenzene, benzene, 1-ethyl-2,4-dimethyl, m-xylene, 4-ethyl, 1,3-dimethyl-4-ethyl benzene, benzene,1-ethyl-2,4-dimethyl, 4-ethyl-1,3-xylene, acmc-1blmc, 2,4-dimethylethylbenzene, 2,4-dimethyl-1-ethylbenzene
PubChem CID 13403
IUPAC Name 1-ethyl-2,4-dimethylbenzene
SMILES CCC1=C(C=C(C=C1)C)C

Specifications

Color Colorless
Quantity 1 mL
Physical Form Clear Liquid at 20°C
UN Number 3295
Boiling Point 186°C
Formula Weight 134.22
Percent Purity ≥95.0% (GC)
Chemical Name or Material 4-Ethyl-m-xylene
EINECSNumber (3)-3427
TSCA Yes