accessibility menu, dialog, popup

UserName

H3CCH3H3C

CAS RN 874-41-9

IUPAC Name: 1-ethyl-2,4-dimethylbenzene
Synonyms: 1,3-dimethyl-4-ethylbenzene
Molecular Weight (g/mol): 134.22
Molecular Formula: C10H14
InChi Key: MEMBJMDZWKVOTB-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C)C=C1C
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)