missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethyl-4'-(trans-4-propylcyclohexyl)biphenyl 98.0+%, TCI America™
$221.39 - $797.02
Chemical Identifiers
| CAS | 84540-37-4 |
|---|---|
| Molecular Formula | C23H30 |
| Molecular Weight (g/mol) | 306.49 |
| MDL Number | MFCD09751090 |
| InChI Key | DOALOUQODWWGEZ-UHFFFAOYSA-N |
| Synonym | 4-ethyl-4'-trans-4-propylcyclohexyl-1,1'-biphenyl, trans-4-ethyl-4'-4-propylcyclohexyl-1,1'-biphenyl, 1-ethyl-4-4-4-propylcyclohexyl phenyl benzene, 4-ethyl-4'-4-propylcyclohexyl biphenyl, 4-ethyl-4'-trans-4-propylcyclohexyl biphenyl, 4-ethyl-4'-4-propylcyclohexyl-1,1'-biphenyl, 1,1'-biphenyl, 4-ethyl-4'-4-propylcyclohexyl, 1,1'-biphenyl, 4-ethyl-4'-trans-4-propylcyclohexyl, acmc-20myl6, 1,1'-biphenyl, 4-ethyl-4'-4-propylcyclohexyl-, trans |
| PubChem CID | 606477 |
| IUPAC Name | 4-ethyl-4'-(4-propylcyclohexyl)-1,1'-biphenyl |
| SMILES | CCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC=C(CC)C=C1 |
Chemical Identifiers
| 84540-37-4 | |
| 306.49 | |
| DOALOUQODWWGEZ-UHFFFAOYSA-N | |
| 606477 | |
| CCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC=C(CC)C=C1 |
| C23H30 | |
| MFCD09751090 | |
| 4-ethyl-4'-trans-4-propylcyclohexyl-1,1'-biphenyl, trans-4-ethyl-4'-4-propylcyclohexyl-1,1'-biphenyl, 1-ethyl-4-4-4-propylcyclohexyl phenyl benzene, 4-ethyl-4'-4-propylcyclohexyl biphenyl, 4-ethyl-4'-trans-4-propylcyclohexyl biphenyl, 4-ethyl-4'-4-propylcyclohexyl-1,1'-biphenyl, 1,1'-biphenyl, 4-ethyl-4'-4-propylcyclohexyl, 1,1'-biphenyl, 4-ethyl-4'-trans-4-propylcyclohexyl, acmc-20myl6, 1,1'-biphenyl, 4-ethyl-4'-4-propylcyclohexyl-, trans | |
| 4-ethyl-4'-(4-propylcyclohexyl)-1,1'-biphenyl |
Specifications
| 84540-37-4 | |
| White | |
| MFCD09751090 | |
| 4-ethyl-4'-trans-4-propylcyclohexyl-1,1'-biphenyl, trans-4-ethyl-4'-4-propylcyclohexyl-1,1'-biphenyl, 1-ethyl-4-4-4-propylcyclohexyl phenyl benzene, 4-ethyl-4'-4-propylcyclohexyl biphenyl, 4-ethyl-4'-trans-4-propylcyclohexyl biphenyl, 4-ethyl-4'-4-propylcyclohexyl-1,1'-biphenyl, 1,1'-biphenyl, 4-ethyl-4'-4-propylcyclohexyl, 1,1'-biphenyl, 4-ethyl-4'-trans-4-propylcyclohexyl, acmc-20myl6, 1,1'-biphenyl, 4-ethyl-4'-4-propylcyclohexyl-, trans | |
| CCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC=C(CC)C=C1 | |
| 306.49 | |
| 306.49 | |
| Crystalline Powder |
| 136°C | |
| C23H30 | |
| 5 g | |
| DOALOUQODWWGEZ-UHFFFAOYSA-N | |
| 4-ethyl-4'-(4-propylcyclohexyl)-1,1'-biphenyl | |
| 606477 | |
| ≥98.0% (GC) | |
| 4-Ethyl-4′-(trans-4-propylcyclohexyl)biphenyl |
Safety and Handling
TSCA : No