missing translation for 'onlineSavingsMsg'
Learn More

4-Ethyl-4'-(trans-4-propylcyclohexyl)biphenyl 98.0+%, TCI America™

Supplier:  TCI America E116225G

 View more versions of this product

Catalog No. E116225G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Ethyl-4′-(trans-4-propylcyclohexyl)biphenyl
136°C
C23H30
25 g
DOALOUQODWWGEZ-UHFFFAOYSA-N
4-ethyl-4'-(4-propylcyclohexyl)-1,1'-biphenyl
606477
≥98.0% (GC)
84540-37-4
White
MFCD09751090
4-ethyl-4'-trans-4-propylcyclohexyl-1,1'-biphenyl, trans-4-ethyl-4'-4-propylcyclohexyl-1,1'-biphenyl, 1-ethyl-4-4-4-propylcyclohexyl phenyl benzene, 4-ethyl-4'-4-propylcyclohexyl biphenyl, 4-ethyl-4'-trans-4-propylcyclohexyl biphenyl, 4-ethyl-4'-4-propylcyclohexyl-1,1'-biphenyl, 1,1'-biphenyl, 4-ethyl-4'-4-propylcyclohexyl, 1,1'-biphenyl, 4-ethyl-4'-trans-4-propylcyclohexyl, acmc-20myl6, 1,1'-biphenyl, 4-ethyl-4'-4-propylcyclohexyl-, trans
CCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC=C(CC)C=C1
306.49
306.49
Crystalline Powder

Chemical Identifiers

84540-37-4
306.49
DOALOUQODWWGEZ-UHFFFAOYSA-N
606477
CCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC=C(CC)C=C1
C23H30
MFCD09751090
4-ethyl-4'-trans-4-propylcyclohexyl-1,1'-biphenyl, trans-4-ethyl-4'-4-propylcyclohexyl-1,1'-biphenyl, 1-ethyl-4-4-4-propylcyclohexyl phenyl benzene, 4-ethyl-4'-4-propylcyclohexyl biphenyl, 4-ethyl-4'-trans-4-propylcyclohexyl biphenyl, 4-ethyl-4'-4-propylcyclohexyl-1,1'-biphenyl, 1,1'-biphenyl, 4-ethyl-4'-4-propylcyclohexyl, 1,1'-biphenyl, 4-ethyl-4'-trans-4-propylcyclohexyl, acmc-20myl6, 1,1'-biphenyl, 4-ethyl-4'-4-propylcyclohexyl-, trans
4-ethyl-4'-(4-propylcyclohexyl)-1,1'-biphenyl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions